Product Details

SNP ID
rs9820928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:123283490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTGCATGTGTGTACATACGTGT[C/G]CGTGTGTCAGAAAAGATGGGGCAGT
Phenotype
MIM: 600293 MIM: 612442
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADCY5 PubMed Links

Gene Details

Gene
ADCY5
Gene Name
adenylate cyclase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199642.1 4000 UTR 3 NP_001186571.1
NM_183357.2 4000 UTR 3 NP_899200.1
XM_005247077.3 4000 UTR 3 XP_005247134.1
XM_005247078.1 4000 UTR 3 XP_005247135.1
XM_006713483.1 4000 UTR 3 XP_006713546.1
XM_006713484.1 4000 UTR 3 XP_006713547.1
XM_011512359.2 4000 UTR 3 XP_011510661.1
XM_011512360.2 4000 UTR 3 XP_011510662.1
XM_011512361.1 4000 UTR 3 XP_011510663.1
XM_017005638.1 4000 UTR 3 XP_016861127.1
XM_017005639.1 4000 UTR 3 XP_016861128.1
XM_017005640.1 4000 UTR 3 XP_016861129.1
Gene
SEC22A
Gene Name
SEC22 homolog A, vesicle trafficking protein
There are no transcripts associated with this gene.

View Full Product Details