Product Details

SNP ID
rs10227405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30968389 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCCCTGTGAGATCTTCAGAAGCA[C/T]ATTGTCTAATGTGTCTCAGAACTTA
Phenotype
MIM: 139191
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GHRHR PubMed Links

Gene Details

Gene
GHRHR
Gene Name
growth hormone releasing hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000823.3 Intron NP_000814.2
XM_011515263.1 Intron XP_011513565.1

View Full Product Details