Product Details

SNP ID
rs9293763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:79072671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACTGCTGTACAGGGGGATGCAT[A/G]AATGAAGGATCTGTGAAGTTAAGCA
Phenotype
MIM: 605932 MIM: 605849
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BHMT2 PubMed Links

Gene Details

Gene
BHMT2
Gene Name
betaine--homocysteine S-methyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178005.1 Intron NP_001171476.1
NM_017614.4 Intron NP_060084.2
Gene
DMGDH
Gene Name
dimethylglycine dehydrogenase
There are no transcripts associated with this gene.

View Full Product Details