Product Details

SNP ID
rs6703976
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:21510441 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGGGCCCTGGGAAAAAGTCAGAC[C/T]GCTCAGGTGGGCCCAGCTGCCCAGG
Phenotype
MIM: 171760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALPL PubMed Links

Gene Details

Gene
ALPL
Gene Name
alkaline phosphatase, liver/bone/kidney
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000478.5 Intron NP_000469.3
NM_001127501.3 Intron NP_001120973.2
NM_001177520.2 Intron NP_001170991.1
XM_005245818.1 Intron XP_005245875.1
XM_006710546.2 Intron XP_006710609.1
XM_017000903.1 Intron XP_016856392.1

View Full Product Details