Product Details

SNP ID
hCV29500076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48511749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGCTCCCCCTGGAGGAGGGAGC[C/G]TGGGAGAATTTTTCCACCATGAGAC
Phenotype
MIM: 300651
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927635 PubMed Links

Gene Details

Gene
LOC101927635
Gene Name
uncharacterized LOC101927635
There are no transcripts associated with this gene.

Gene
PORCN
Gene Name
porcupine homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282167.1 Intron NP_001269096.1
NM_022825.3 Intron NP_073736.2
NM_203473.2 Intron NP_982299.1
NM_203474.1 Intron NP_982300.1
NM_203475.2 Intron NP_982301.1

View Full Product Details