Product Details

SNP ID
rs10154668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46244213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGGGGGTGCTGCAGGAACCCCC[A/G]GCAGGCAGTGGGGCCAGGCTTCACA
Phenotype
MIM: 170998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDPF1 PubMed Links

Gene Details

Gene
CDPF1
Gene Name
cysteine rich DPF motif domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207327.4 1679 UTR 3 NP_997210.3
XM_011529960.1 1679 UTR 3 XP_011528262.1
XM_011529962.2 1679 UTR 3 XP_011528264.1
XM_011529963.1 1679 Intron XP_011528265.1
XM_011529964.1 1679 Intron XP_011528266.1
XM_011529965.2 1679 UTR 3 XP_011528267.1
Gene
PPARA
Gene Name
peroxisome proliferator activated receptor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001928.2 1679 Intron NP_001001928.1
NM_005036.4 1679 Intron NP_005027.2
XM_005261655.3 1679 Intron XP_005261712.1
XM_005261656.3 1679 Intron XP_005261713.1
XM_006724269.3 1679 Intron XP_006724332.1
XM_006724270.3 1679 Intron XP_006724333.1
XM_011530239.2 1679 Intron XP_011528541.1
XM_011530240.2 1679 Intron XP_011528542.1
XM_011530241.2 1679 Intron XP_011528543.1
XM_011530242.2 1679 Intron XP_011528544.1
XM_011530243.2 1679 Intron XP_011528545.1
XM_011530244.2 1679 Intron XP_011528546.1
XM_011530245.2 1679 Intron XP_011528547.1
XM_017028839.1 1679 Intron XP_016884328.1
XM_017028840.1 1679 Intron XP_016884329.1

View Full Product Details