Product Details

SNP ID
rs9498148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:148753317 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCATCAACCCCAGTCCTAAGCAAC[C/T]GCTAATTTACTTTCTGTCTGTATAG
Phenotype
MIM: 610752
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UST PubMed Links

Gene Details

Gene
UST
Gene Name
uronyl 2-sulfotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005715.2 Intron NP_005706.1
XM_011535378.2 Intron XP_011533680.1
XM_017010152.1 Intron XP_016865641.1

View Full Product Details