Product Details

SNP ID
rs6005982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29080278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTATATCCTTTCAATCAAATTA[A/G]AGACTGTAGCATCTGGTGAGGGCTG
Phenotype
MIM: 609898
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C22orf31 PubMed Links

Gene Details

Gene
C22orf31
Gene Name
chromosome 22 open reading frame 31
There are no transcripts associated with this gene.

Gene
KREMEN1
Gene Name
kringle containing transmembrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039570.2 Intron NP_001034659.2
NM_032045.4 Intron NP_114434.3

View Full Product Details