Product Details

SNP ID
rs9939975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20783435 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTATACTGGTTGGGCTGCCCTAA[C/T]CCAAAAACATGAAACCTGAAATCCT
Phenotype
MIM: 145505
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACSM3 PubMed Links

Gene Details

Gene
ACSM3
Gene Name
acyl-CoA synthetase medium-chain family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005622.3 Intron NP_005613.2
NM_202000.2 Intron NP_973729.1
XM_011545911.1 Intron XP_011544213.1
XM_017023523.1 Intron XP_016879012.1
XM_017023524.1 Intron XP_016879013.1
Gene
ERI2
Gene Name
ERI1 exoribonuclease family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142725.1 Intron NP_001136197.1
NM_080663.2 Intron NP_542394.2

View Full Product Details