Product Details

SNP ID
rs9928915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4811546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGACTGGGGAGGGGCATCTTTCA[C/T]GCTCACTAAATCCCTGACCTAGTAC
Phenotype
MIM: 610660 MIM: 614574
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLYR1 PubMed Links

Gene Details

Gene
GLYR1
Gene Name
glyoxylate reductase 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308096.1 Intron NP_001295025.1
NM_001324096.1 Intron NP_001311025.1
NM_001324097.1 Intron NP_001311026.1
NM_001324098.1 Intron NP_001311027.1
NM_032569.3 Intron NP_115958.2
XM_005255639.4 Intron XP_005255696.2
XM_011522716.2 Intron XP_011521018.1
XM_011522717.1 Intron XP_011521019.1
XM_017023786.1 Intron XP_016879275.1
XM_017023787.1 Intron XP_016879276.1
XM_017023788.1 Intron XP_016879277.1
XM_017023789.1 Intron XP_016879278.1
Gene
ROGDI
Gene Name
rogdi homolog
There are no transcripts associated with this gene.

View Full Product Details