Product Details

SNP ID
rs6062129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62187142 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGACGGTGGTACCACCTGTTGATA[C/T]AGTGAAGTGCCTGGGCTGAGTTTTG
Phenotype
MIM: 610919 MIM: 606472
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTG2 PubMed Links

Gene Details

Gene
MTG2
Gene Name
mitochondrial ribosome associated GTPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015666.3 Intron NP_056481.1
XM_005260393.1 Intron XP_005260450.1
XM_006723774.3 Intron XP_006723837.1
XM_011528776.1 Intron XP_011527078.1
XM_017027805.1 Intron XP_016883294.1
XM_017027806.1 Intron XP_016883295.1
XM_017027807.1 Intron XP_016883296.1
XM_017027808.1 Intron XP_016883297.1
Gene
SS18L1
Gene Name
SS18L1, nBAF chromatin remodeling complex subunit
There are no transcripts associated with this gene.

View Full Product Details