Product Details

SNP ID
rs6503311
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:10444226 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAATGAAAAAGATAACTGCAGCC[A/G]AATTGAAAAAAAAAGTCCCAAGATG
Phenotype
MIM: 160742
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MYH4 PubMed Links

Gene Details

Gene
MYH4
Gene Name
myosin heavy chain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017533.2 Intron NP_060003.2
XM_017024676.1 Intron XP_016880165.1
Gene
MYHAS
Gene Name
myosin heavy chain gene cluster antisense RNA
There are no transcripts associated with this gene.

View Full Product Details