Product Details

SNP ID
rs10042821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:95720915 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGTCATTTGTTGCCATATTTTAC[C/T]GAAGAAAAGGATTTTCTGGCTCTTT
Phenotype
MIM: 607353 MIM: 608039
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RHOBTB3 PubMed Links

Gene Details

Gene
RHOBTB3
Gene Name
Rho related BTB domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014899.3 Intron NP_055714.3
XM_011543279.2 Intron XP_011541581.1
XM_017009236.1 Intron XP_016864725.1
XM_017009237.1 Intron XP_016864726.1
Gene
SPATA9
Gene Name
spermatogenesis associated 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031952.3 Intron NP_114158.2
XM_011543664.1 Intron XP_011541966.1
XM_011543666.1 Intron XP_011541968.1
XM_011543667.2 Intron XP_011541969.1
XM_017009948.1 Intron XP_016865437.1
XM_017009949.1 Intron XP_016865438.1
XM_017009950.1 Intron XP_016865439.1
XM_017009951.1 Intron XP_016865440.1

View Full Product Details