Product Details

SNP ID
rs10403016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46875454 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTGAATGAGAGAATCCTGGAATA[C/G]ATGAAGTCTGTCGCCTTTCAGTCTT
Phenotype
MIM: 605277
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP35 PubMed Links

Gene Details

Gene
ARHGAP35
Gene Name
Rho GTPase activating protein 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004491.4 Intron NP_004482.4
XM_011526873.2 Intron XP_011525175.1
XM_017026714.1 Intron XP_016882203.1

View Full Product Details