Product Details

SNP ID
rs1048087
Assay Type
Functionally Tested
NCBI dbSNP Submissions
10
Location
Chr.6:32641509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAAACATGGCTGTGGCAAAACA[C/T]AACTTGAACATCATGATTAAACGCT
Phenotype
MIM: 146880
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HLA-DQA1 PubMed Links

Gene Details

Gene
HLA-DQA1
Gene Name
major histocompatibility complex, class II, DQ alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002122.3 338 Silent Mutation CAC,CAT H94H NP_002113.2
XM_006715079.3 338 Silent Mutation CAC,CAT H94H XP_006715142.1

View Full Product Details