Product Details

SNP ID
rs3118417
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:68428954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAAGTGATTGCACAATGCTTAAA[C/T]ACAATTAAACAAAAAAGATGCAATA
Phenotype
MIM: 180069
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPE65 PubMed Links

Gene Details

Gene
RPE65
Gene Name
RPE65, retinoid isomerohydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000329.2 2690 UTR 3 NP_000320.1
XM_017002027.1 2690 UTR 3 XP_016857516.1

View Full Product Details