Product Details

SNP ID
rs10475010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:14154333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTTCTAACTTAGGAAAGGAAC[A/G]GGGCCGGGGAGTGAGAGGGGCAGGA
Phenotype
MIM: 601893
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIO PubMed Links

Gene Details

Gene
TRIO
Gene Name
trio Rho guanine nucleotide exchange factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007118.3 Intron NP_009049.2
XM_011514107.2 Intron XP_011512409.1
XM_011514108.1 Intron XP_011512410.1
XM_011514109.2 Intron XP_011512411.1
XM_011514110.2 Intron XP_011512412.1
XM_017009801.1 Intron XP_016865290.1
XM_017009802.1 Intron XP_016865291.1
XM_017009803.1 Intron XP_016865292.1

View Full Product Details