Product Details

SNP ID
rs6031989
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45094446 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCAACAAGCCCAGATTCCAAAC[C/T]TGACTCTGCCACTTCTAATTGTGAC
Phenotype
MIM: 602905
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KCNS1 PubMed Links

Gene Details

Gene
KCNS1
Gene Name
potassium voltage-gated channel modifier subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322799.1 2402 UTR 3 NP_001309728.1
NM_002251.4 2402 UTR 3 NP_002242.2
XM_017027846.1 2402 UTR 3 XP_016883335.1

View Full Product Details