Product Details

SNP ID
rs6142998
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62228407 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCGGGAAGAGGCGCCACGTGGAGG[A/G]GCTGAGCAGTGCAGGTGGGCTCAGG
Phenotype
MIM: 604525
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HRH3 PubMed Links

Gene Details

Gene
HRH3
Gene Name
histamine receptor H3
There are no transcripts associated with this gene.

Gene
LOC105369209
Gene Name
putative uncharacterized protein FLJ44790
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011529124.1 Intron XP_011527426.1

View Full Product Details