Product Details
- SNP ID
-
rs9834044
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:46830861 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCATATTTTCAGTTCCACTCTCA[A/G]ACAGCCCCACACCCTCCAACTCAAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRSS42
PubMed Links
Gene Details
- Gene
- PRSS42
- Gene Name
- protease, serine 42
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182702.1 |
|
Intron |
|
|
NP_874361.1 |
View Full Product Details