Product Details

SNP ID
rs10026933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70903213 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCAGAGGACTTCCCTGAAGAGGA[C/T]GAGTTTTATCTTTTTAGACAACCAG
Phenotype
MIM: 609282
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MOB1B PubMed Links

Gene Details

Gene
MOB1B
Gene Name
MOB kinase activator 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244766.1 Intron NP_001231695.1
NM_001244767.1 Intron NP_001231696.1
NM_173468.3 Intron NP_775739.1
XM_005265709.2 Intron XP_005265766.1
XM_011532412.2 Intron XP_011530714.1
XM_017008837.1 Intron XP_016864326.1

View Full Product Details