Product Details

SNP ID
rs4789146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75139498 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAATAAAAAAGCACCTATGGAGA[C/T]TAAACAAGCAGCAGTAGGCCTTACT
Phenotype
MIM: 191720
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARMC7 PubMed Links

Gene Details

Gene
ARMC7
Gene Name
armadillo repeat containing 7
There are no transcripts associated with this gene.

Gene
HN1
Gene Name
hematological and neurological expressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002032.2 Intron NP_001002032.1
NM_001002033.2 Intron NP_001002033.1
NM_001288609.1 Intron NP_001275538.1
NM_001288610.1 Intron NP_001275539.1
NM_001288611.1 Intron NP_001275540.1
NM_016185.3 Intron NP_057269.1
Gene
NT5C
Gene Name
5', 3'-nucleotidase, cytosolic
There are no transcripts associated with this gene.

View Full Product Details