Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001195248.1 | 3311 | Intron | NP_001182177.1 | ||
NM_001195249.1 | 3311 | Intron | NP_001182178.1 | ||
NM_001195250.1 | 3311 | Intron | NP_001182179.1 | ||
NM_001195251.1 | 3311 | Intron | NP_001182180.1 | ||
NM_001195252.1 | 3311 | Intron | NP_001182181.1 | ||
NM_001195254.1 | 3311 | Intron | NP_001182183.1 | ||
NM_175069.2 | 3311 | Intron | NP_778239.1 | ||
NM_175073.2 | 3311 | Intron | NP_778243.1 | ||
XM_006716791.3 | 3311 | Intron | XP_006716854.1 | ||
XM_006716792.2 | 3311 | Intron | XP_006716855.1 | ||
XM_011517938.1 | 3311 | Intron | XP_011516240.1 | ||
XM_011517939.2 | 3311 | Intron | XP_011516241.1 | ||
XM_017014831.1 | 3311 | UTR 3 | XP_016870320.1 | ||
XM_017014832.1 | 3311 | Intron | XP_016870321.1 | ||
XM_017014833.1 | 3311 | Intron | XP_016870322.1 | ||
XM_017014834.1 | 3311 | Intron | XP_016870323.1 | ||
XM_017014835.1 | 3311 | Intron | XP_016870324.1 | ||
XM_017014836.1 | 3311 | Intron | XP_016870325.1 | ||
XM_017014837.1 | 3311 | Intron | XP_016870326.1 | ||
XM_017014838.1 | 3311 | Intron | XP_016870327.1 |