Product Details

SNP ID
rs9426420
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:28567060 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCGAGGAGGATGGTGAACTGAAT[A/G]TGGGAATTCCTGCAGATATTAGAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRNAU1AP PubMed Links

Gene Details

Gene
TRNAU1AP
Gene Name
tRNA selenocysteine 1 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017846.4 Intron NP_060316.1
XM_017001542.1 Intron XP_016857031.1

View Full Product Details