Product Details

SNP ID
rs10178810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:196195478 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATTCTGCTGAGCTGTTCAATTAC[A/G]AAAGCCAAATAAACGACTCTGCAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HECW2 PubMed Links

Gene Details

Gene
HECW2
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304840.1 10738 Intron NP_001291769.1
NM_020760.2 10738 Intron NP_065811.1
XM_006712646.3 10738 UTR 3 XP_006712709.1
XM_006712648.3 10738 UTR 3 XP_006712711.1
XM_017004563.1 10738 UTR 3 XP_016860052.1

View Full Product Details