Product Details

SNP ID
rs10203981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96276393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATGGACAATACTACACAACTGA[C/G]GGGTTTAACAATGACCAGTCATCTA
Phenotype
MIM: 604333 MIM: 601664
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CIAO1 PubMed Links

Gene Details

Gene
CIAO1
Gene Name
cytosolic iron-sulfur assembly component 1
There are no transcripts associated with this gene.

Gene
SNRNP200
Gene Name
small nuclear ribonucleoprotein U5 subunit 200
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014014.4 Intron NP_054733.2
XM_017003603.1 Intron XP_016859092.1

View Full Product Details