Product Details

SNP ID
rs6642304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:155615387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGAAGCTTTAGTGATCTCTAATC[A/G]TATTCATAACCTTGAGTTCTTGAAA
Phenotype
MIM: 300531 MIM: 300777
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPRY3 PubMed Links

Gene Details

Gene
SPRY3
Gene Name
sprouty RTK signaling antagonist 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304990.1 Intron NP_001291919.1
NM_005840.2 Intron NP_005831.1
Gene
TMLHE
Gene Name
trimethyllysine hydroxylase, epsilon
There are no transcripts associated with this gene.

View Full Product Details