Product Details

SNP ID
rs9827980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:115628302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTGAGATTTCATCTTCTTGTGC[C/T]GCACTGGCTCCTTTCCTTATAGTTT
Phenotype
MIM: 162060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAP43 PubMed Links

Gene Details

Gene
GAP43
Gene Name
growth associated protein 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130064.1 Intron NP_001123536.1
NM_002045.3 Intron NP_002036.1
XM_017006128.1 Intron XP_016861617.1

View Full Product Details