Product Details

SNP ID
rs9263804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31167929 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATAGTGACTTTGTTTTAGTCCAAG[C/T]GAAATGATCTCTCACCCTTTTTCTC
Phenotype
MIM: 605310 MIM: 164177 MIM: 600912
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCHCR1 PubMed Links

Gene Details

Gene
CCHCR1
Gene Name
coiled-coil alpha-helical rod protein 1
There are no transcripts associated with this gene.

Gene
POU5F1
Gene Name
POU class 5 homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173531.2 Intron NP_001167002.1
NM_001285986.1 Intron NP_001272915.1
NM_001285987.1 Intron NP_001272916.1
NM_002701.5 Intron NP_002692.2
NM_203289.5 Intron NP_976034.4
Gene
PSORS1C3
Gene Name
psoriasis susceptibility 1 candidate 3 (non-protein coding)
There are no transcripts associated with this gene.

Gene
TCF19
Gene Name
transcription factor 19
There are no transcripts associated with this gene.

View Full Product Details