Product Details

SNP ID
rs6413526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45485060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGAGACTGATGCTTGATACGGA[C/T]ACAGCTGTCAGGGATTCCATTTTCT
Phenotype
MIM: 602095
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105376714 PubMed Links

Gene Details

Gene
LOC105376714
Gene Name
uncharacterized LOC105376714
There are no transcripts associated with this gene.

Gene
SLC30A4
Gene Name
solute carrier family 30 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321036.1 1511 Intron NP_001307965.1
NM_013309.5 1511 UTR 3 NP_037441.2
XM_011521997.2 1511 UTR 3 XP_011520299.1
XM_017022560.1 1511 UTR 3 XP_016878049.1
XM_017022561.1 1511 Intron XP_016878050.1
XM_017022562.1 1511 Intron XP_016878051.1

View Full Product Details