Product Details

SNP ID
rs5906352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47195882 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATGTCAGATACACCTCTCCTCTT[A/G]CATGTCCCTATCTATTATTTTTGGT
Phenotype
MIM: 300164 MIM: 300080 MIM: 314370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INE1 PubMed Links

Gene Details

Gene
INE1
Gene Name
inactivation escape 1 (non-protein coding)
There are no transcripts associated with this gene.

Gene
RBM10
Gene Name
RNA binding motif protein 10
There are no transcripts associated with this gene.

Gene
UBA1
Gene Name
ubiquitin like modifier activating enzyme 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003334.3 1858 Intron NP_003325.2
NM_153280.2 1858 Intron NP_695012.1
XM_005272649.1 1858 Intron XP_005272706.1
XM_011543954.2 1858 Intron XP_011542256.1
XM_017029777.1 1858 Intron XP_016885266.1
XM_017029778.1 1858 UTR 5 XP_016885267.1
XM_017029779.1 1858 UTR 5 XP_016885268.1
XM_017029780.1 1858 Intron XP_016885269.1
XM_017029781.1 1858 Intron XP_016885270.1

View Full Product Details