Product Details

SNP ID
rs7157625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:58422656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTCTTTTCAGTGAAATAAAGT[C/T]TGTCATACTATAAAAGTAATATATT
Phenotype
MIM: 610178 MIM: 607384
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA0586 PubMed Links

Gene Details

Gene
KIAA0586
Gene Name
KIAA0586
There are no transcripts associated with this gene.

Gene
TIMM9
Gene Name
translocase of inner mitochondrial membrane 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304485.1 Intron NP_001291414.1
NM_001304486.1 Intron NP_001291415.1
NM_001304487.1 Intron NP_001291416.1
NM_001304488.1 Intron NP_001291417.1
NM_001304489.1 Intron NP_001291418.1
NM_001304490.1 Intron NP_001291419.1
NM_001304491.1 Intron NP_001291420.1
NM_012460.3 Intron NP_036592.1
Gene
TOMM20L
Gene Name
translocase of outer mitochondrial membrane 20 like
There are no transcripts associated with this gene.

View Full Product Details