Product Details

SNP ID
rs9697674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123485823 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTCCGTCTGGAGGAGGCAGAGAT[A/G]CTGCTAATGAATTCTTGGCAGCCAC
Phenotype
MIM: 614092
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RILPL1 PubMed Links

Gene Details

Gene
RILPL1
Gene Name
Rab interacting lysosomal protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319243.1 Intron NP_001306172.1
NM_001319244.1 Intron NP_001306173.1
NM_001319302.1 Intron NP_001306231.1
NM_178314.4 Intron NP_847884.2
XM_006719372.2 Intron XP_006719435.1
XM_006719373.3 Intron XP_006719436.1
XM_011538282.2 Intron XP_011536584.1
XM_011538284.2 Intron XP_011536586.1
XM_011538285.2 Intron XP_011536587.1

View Full Product Details