Product Details
- SNP ID
-
rs9914194
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40629466 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATTTCTGTTGGACAGTGAATGAGA[A/G]TTAGTCTGTAGTGCTATGGATTAAA
- Phenotype
-
MIM: 603111
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SMARCE1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77380256] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SMARCE1
- Gene Name
- SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003079.4 |
|
Intron |
|
|
NP_003070.3 |
View Full Product Details