Product Details

SNP ID
rs6002123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40774712 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAACATCTGCAGTCTAAGCTCTTCA[C/G]ATTATGTGTTTACATTATAAAAGTG
Phenotype
MIM: 606795
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A17 PubMed Links

Gene Details

Gene
SLC25A17
Gene Name
solute carrier family 25 member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282726.1 Intron NP_001269655.1
NM_001282727.1 Intron NP_001269656.1
NM_006358.3 Intron NP_006349.1

View Full Product Details