Product Details

SNP ID
rs7554146
Assay Type
Functionally Tested
NCBI dbSNP Submissions
25
Location
Chr.1:147544859 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCTCTCTCTCTCTCTCTTCCTC[C/T]TATTTGTGGCCTTTCTTGGAAAGAA
Phenotype
MIM: 602597
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
BCL9 PubMed Links

Gene Details

Gene
BCL9
Gene Name
B-cell CLL/lymphoma 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004326.3 Intron NP_004317.2
XM_005272971.4 Intron XP_005273028.1
XM_005277417.3 Intron XP_005277474.1
XM_005277418.3 Intron XP_005277475.1
XM_017002026.1 Intron XP_016857515.1

View Full Product Details