Product Details
- SNP ID
-
rs9831052
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:112121257 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCTACTTCCACAATGGTCCAGTGG[C/T]CATGTGAAAAATTCTAGCATAAACT
- Phenotype
-
MIM: 611956
MIM: 607792
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C3orf52
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114214934] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C3orf52
- Gene Name
- chromosome 3 open reading frame 52
- Gene
- GCSAM
- Gene Name
- germinal center associated signaling and motility
- Gene
- MIR567
- Gene Name
- microRNA 567
There are no transcripts associated with this gene.
View Full Product Details