Product Details

SNP ID
rs9611179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39452127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGAGAAATTGCAAGCATTTCA[A/G]AGGGCTGCCAGTGGATGGGACAAGC
Phenotype
MIM: 604621
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MGAT3 PubMed Links

Gene Details

Gene
MGAT3
Gene Name
mannosyl (beta-1,4-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098270.1 Intron NP_001091740.1
NM_002409.4 Intron NP_002400.3
XM_005261608.4 Intron XP_005261665.1
XM_005261609.2 Intron XP_005261666.1
XM_006724252.2 Intron XP_006724315.1
XM_011530184.2 Intron XP_011528486.1
XM_011530185.2 Intron XP_011528487.1
XM_011530186.2 Intron XP_011528488.1
XM_011530187.2 Intron XP_011528489.1
XM_011530188.2 Intron XP_011528490.1
XM_011530189.2 Intron XP_011528491.1
XM_011530190.2 Intron XP_011528492.1
XM_011530191.2 Intron XP_011528493.1
XM_011530192.2 Intron XP_011528494.1
XM_011530194.2 Intron XP_011528496.1
XM_017028802.1 Intron XP_016884291.1

View Full Product Details