Product Details

SNP ID
rs104894297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:121303512 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAATGTTGGTGCTTACATCCTTT[A/C]TTTCTTCTGTGCAACACTGAGCTAT
Phenotype
MIM: 602286
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SC5D PubMed Links

Gene Details

Gene
SC5D
Gene Name
sterol-C5-desaturase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024956.2 358 Missense Mutation TAT,TCT Y46S NP_001020127.1
NM_006918.4 358 Missense Mutation TAT,TCT Y46S NP_008849.2

View Full Product Details