Product Details

SNP ID
rs9921905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:85712672 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCTCAGAAAAACTGAAAGCAGTA[C/G]GCACACTGGCTTTGGAATCAGGCCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C16orf74 PubMed Links

Gene Details

Gene
C16orf74
Gene Name
chromosome 16 open reading frame 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206967.2 Intron NP_996850.1

View Full Product Details