Product Details
- SNP ID
-
rs6137954
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23452601 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACACTCACCTGCCTCTGGACTTTAA[A/G]GACTCGGAAGATCCTGAAGTGGTAC
- Phenotype
-
MIM: 609731
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CST11
PubMed Links
Gene Details
- Gene
- CST11
- Gene Name
- cystatin 11
- Gene
- CSTL1
- Gene Name
- cystatin like 1
There are no transcripts associated with this gene.
View Full Product Details