Product Details
- SNP ID
-
rs9468892
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:31202114 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATGTACACCTATGCTCTGTACATA[A/C]CCAGGGCTCTGCACTGAGGCAGACC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HCG27
PubMed Links
Gene Details
- Gene
- HCG27
- Gene Name
- HLA complex group 27 (non-protein coding)
There are no transcripts associated with this gene.
View Full Product Details