Product Details
- SNP ID
-
rs6597087
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:4706428 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTGTAACTTTAAGAGTTTGGGTG[G/T]TTTTTTTTTCTTTTCATTTGTTTGT
- Phenotype
-
MIM: 603778
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CDYL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114726049] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDYL
- Gene Name
- chromodomain protein, Y-like
View Full Product Details