Product Details

SNP ID
rs10521527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:110202764 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCCCAACATGCACTAGGCTATGA[C/T]ACTATGTTTGGAAAACTGGAACAAC
Phenotype
MIM: 300195
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMMECR1 PubMed Links

Gene Details

Gene
AMMECR1
Gene Name
Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025580.1 Intron NP_001020751.1
NM_001171689.1 Intron NP_001165160.1
NM_015365.2 Intron NP_056180.1

View Full Product Details