Product Details

SNP ID
rs9850496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:179796922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGATACCCCACACACTTGCTAT[A/G]TGACATTTATGAAAATTAAGGCCCC
Phenotype
MIM: 611058 MIM: 603591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PEX5L PubMed Links

Gene Details

Gene
PEX5L
Gene Name
peroxisomal biogenesis factor 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256750.1 8435 UTR 3 NP_001243679.1
NM_001256751.1 8435 UTR 3 NP_001243680.1
NM_001256752.1 8435 UTR 3 NP_001243681.1
NM_001256753.1 8435 UTR 3 NP_001243682.1
NM_001256754.1 8435 UTR 3 NP_001243683.1
NM_001256755.1 8435 UTR 3 NP_001243684.1
NM_001256756.1 8435 UTR 3 NP_001243685.1
NM_016559.2 8435 UTR 3 NP_057643.1
XM_011512882.2 8435 UTR 3 XP_011511184.1
XM_011512884.2 8435 UTR 3 XP_011511186.1
XM_011512885.2 8435 UTR 3 XP_011511187.1
XM_011512886.2 8435 UTR 3 XP_011511188.1
XM_011512887.2 8435 UTR 3 XP_011511189.1
XM_011512888.2 8435 UTR 3 XP_011511190.1
XM_011512891.2 8435 UTR 3 XP_011511193.1
XM_011512892.2 8435 UTR 3 XP_011511194.1
XM_017006601.1 8435 UTR 3 XP_016862090.1
XM_017006602.1 8435 UTR 3 XP_016862091.1
XM_017006603.1 8435 UTR 3 XP_016862092.1
XM_017006604.1 8435 UTR 3 XP_016862093.1
XM_017006605.1 8435 UTR 3 XP_016862094.1
XM_017006606.1 8435 UTR 3 XP_016862095.1
XM_017006607.1 8435 UTR 3 XP_016862096.1
XM_017006608.1 8435 UTR 3 XP_016862097.1
XM_017006609.1 8435 UTR 3 XP_016862098.1
XM_017006610.1 8435 UTR 3 XP_016862099.1
XM_017006611.1 8435 UTR 3 XP_016862100.1
Gene
USP13
Gene Name
ubiquitin specific peptidase 13 (isopeptidase T-3)
There are no transcripts associated with this gene.

View Full Product Details