Product Details

SNP ID
rs9306109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44637673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTAGCTGCCAGCCGGATTGCTGC[A/T]CCTCCTCCCCCTGCCAGCAGGCCTG
Phenotype
MIM: 612920
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP10-10 PubMed Links

Gene Details

Gene
KRTAP10-10
Gene Name
keratin associated protein 10-10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181688.2 Intron NP_859016.1
Gene
KRTAP10-11
Gene Name
keratin associated protein 10-11
There are no transcripts associated with this gene.

Gene
KRTAP10-9
Gene Name
keratin associated protein 10-9
There are no transcripts associated with this gene.

Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272037.1 Intron NP_001258966.1
NM_144991.2 Intron NP_659428.2

View Full Product Details