Product Details

SNP ID
rs6641316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:149477528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGACTATCACCTAGAATCCAGA[A/G]TCAAACAAGGGCATCCTTTGGTGAT
Phenotype
MIM: 300823
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IDS PubMed Links

Gene Details

Gene
IDS
Gene Name
iduronate 2-sulfatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000202.7 7044 UTR 3 NP_000193.1
NM_001166550.3 7044 UTR 3 NP_001160022.1
NM_006123.4 7044 Intron NP_006114.1

View Full Product Details