Product Details
- SNP ID
-
rs9306388
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:23645769 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGCCTATTTTTAAATAATACTT[A/T]ATTTTCTTTCATTTTATTTGCTAAT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DRICH1
PubMed Links
Gene Details
- Gene
- DRICH1
- Gene Name
- aspartate rich 1
There are no transcripts associated with this gene.
- Gene
- GUSBP11
- Gene Name
- glucuronidase, beta pseudogene 11
There are no transcripts associated with this gene.
View Full Product Details