Product Details

SNP ID
rs6121292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31949077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGTTGTAGAGAAAAATAAAGCA[A/G]GCTAAGATGGCTAGGGAATGCCAGA
Phenotype
MIM: 610789
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDRG1 PubMed Links

Gene Details

Gene
PDRG1
Gene Name
p53 and DNA damage regulated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030815.2 Intron NP_110442.1
Gene
TTLL9
Gene Name
tubulin tyrosine ligase like 9
There are no transcripts associated with this gene.

View Full Product Details