Product Details

SNP ID
rs6700342
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:212863091 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATATATAATTTACAAATTTTTCT[C/G]ATTCTGTAGGTCATCTTTTGTCAGC
Phenotype
MIM: 609144 MIM: 610864
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FLVCR1 PubMed Links

Gene Details

Gene
FLVCR1
Gene Name
feline leukemia virus subgroup C cellular receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014053.3 Intron NP_054772.1
XM_011509446.2 Intron XP_011507748.1
XM_011509447.2 Intron XP_011507749.1
XM_011509448.1 Intron XP_011507750.1
Gene
FLVCR1-AS1
Gene Name
FLVCR1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details